Learn More
L-Anserine-d4 (N-beta-alanyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | L-Anserine-d4 (N-beta-alanyl-d4) |
| CAS | 1201658-81-2 |
| Molecular Formula | C10 D4 H12 N4 O3 |
| Quantity | 0.01 g |
| Synonym | L-Anserine-d4 (N-β-alanyl-d4), L-Histidine, β-alanyl-2,2,3,3-d4-3-methyl- (ACI), β-Alanyl-2,2,3,3-d4-3-methyl-L-histidine (ACI) |
| InChI Formula | InChI=1 S/C10H16N4O3/c1-14-6-12-5-7(14)4-8(10(16)17)13-9(15)2-3-11/h5-6,8 H,2-4,11H2,1H3,(H,13,15)(H,16,17)/t8-/m0/s1/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])C(=O)N[C@@H](Cc1cncn1C)C(=O)O |
| IUPAC Name | (2 S)-2-[(3-amino-2,2,3,3-tetradeuteriopropanoyl)amino]-3-(3-methylimidazol-4-yl)propanoic acid |
| Molecular Weight (g/mol) | 244.29 |
| Formula Weight | 244.1473 g/mol |
| Show More |