Learn More
Melford L-Asparagine Anhydrous
Description
L-Asparagine Anhydrous is commonly used as a component in cell culture media.
L-Asparagine Anhydrous is commonly used as a component in cell culture media. It provides a vital nitrogen source for cell growth and is particularly important in the cultivation of certain cell types, especially in the production of recombinant proteins and antibodies.
L-Asparagine Anhydrous is utilized in laboratory research, particularly in studies related to cell biology, molecular biology, and biochemistry. Its role as an amino acid makes it valuable for understanding cellular processes.
Specifications
Specifications
| Chemical Name or Material | L-Asparagine, Anhydrous |
| CAS | 70-47-3 |
| Molecular Formula | C4H8N2O3 |
| Quantity | 100 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.