Learn More
L-Aspartic acid 1-benzyl ester, 98%, Thermo Scientific™
Catalog Number p-7054730
Click to view available options
Quantity:
1 g
5 g
7362-93-8
C11H13NO4
223.228
MFCD00063186
NJSRYBIBUXBNSW-VIFPVBQESA-N
h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
6994684
(3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
7362-93-8
C11H13NO4
223.228
MFCD00063186
NJSRYBIBUXBNSW-VIFPVBQESA-N
h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
6994684
(3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Aspartic acid 1-benzyl ester, is used as an intermediate in organic synthesis. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in HCI solution.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Beilstein 2506965
Solubility Information Soluble in HCI solution.
Formula Weight 223.23
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material L-Aspartic acid 1-benzyl ester