missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid, TRC

Product Code. 17938924
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17938924 10 g 10g
30138025 100 g 100g
30132747 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938924 Supplier Toronto Research Chemicals Supplier No. TRCA79002410G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-84-8
Formula Weight 133.0375
InChI Formula InChI=1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1
IUPAC Name (2S)-2-aminobutanedioic acid
Molecular Formula C4 H7 N O4
Molecular Weight (g/mol) 133.1
Percent Purity >95%
SMILES N[C@@H](CC(=O)O)C(=O)O
Synonym Aspartic acid, L- (8CI), (+)-Aspartic acid, (+)-L-Aspartic acid, (S)-2-Aminobutanedioic acid, (S)-Aminobutanedioic acid, (S)-Aspartic acid, 3: PN: WO2018027024 SEQID: 3 claimed protein, 74: PN: WO2018075807 SEQID: 166 claimed protein, Asparagic acid, Asparaginic acid, Aspartic acid, Butanedioic acid, amino-, (S)-, H-Asp-OH, L-(+)-Aspartic acid, L-Aminosuccinic acid, L-Asparagic acid, L-Asparaginic acid, NSC 3973, NSC 79553
Chemical Name or Material L-Aspartic acid
Grade HPLC
Show More Show Less

Specifications

Analyte or Component Names L-Aspartic acid
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.