Learn More
Tocris Bioscience™ L-Azidohomoalanine hydrochloride
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
L-Azidohomoalanine hydrochloride is an unnatural amino acid. Incorporated into proteins during de novo protein synthesis, with no effect on protein degradation. Requires methionine-free medium for efficient incorporation. Substrate for wild-type methionyl-tRNA synthetase (MetRS) in vivo. Can be used in BONCAT (bio-orthogonal non-canonical amino acid tagging) for the identification of newly synthesized proteins.
Specifications
Specifications
| Chemical Name or Material | (S)-2-Amino-4-azidobutanoic acid hydrochloride |
| CAS | 942518-29-8 |
| Quantity | 10 mg |
| Molecular Formula | C4H8N4O2.HCl |
| Formula Weight | 180.59 |