missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L-Ficolin Monoclonal Antibody (GN4), Biotin
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549045
This item is not returnable.
View return policy
Description
L-Ficolin Monoclonal Antibody for IHC (F), IP, IA
L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
Specifications
| L-Ficolin | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Q15485 | |
| FCN2 | |
| The monoclonal antibody GN4 recognizes human Ficolin-2, also known as L-ficolin. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Inhibition Assays, Immunohistochemistry (Frozen), Immunoprecipitation | |
| GN4 | |
| Biotin | |
| FCN2 | |
| 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2220 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction