missing translation for 'onlineSavingsMsg'
Learn More

L-Histidyl-D-tryptophyl-L-alanyl-L-tryptophyl-D-phenylalanyl-L-lysinamide Acetate Salt, TRC

Product Code. 30011380
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011380 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30011380 Supplier Toronto Research Chemicals Supplier No. TRCH4562101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 87616-84-0
Molecular Formula C46H56N12O6. x(C2H4O2)
Synonym L-Lysinamide, L-histidyl-D-tryptophyl-L-alanyl-L-tryptophyl-D-phenylalanyl- (9CI, ACI), L-Histidyl-D-tryptophyl-L-alanyl-L-tryptophyl-D-phenylalanyl-L-lysinamide (ACI), Growth hormone-releasing peptide 6, Hexapeptide 2, His-D-Trp-Ala-Trp-D-Phe-Lys-NH2, SKF 110679, U 75799E, GHRP-6 (His-D-Trp-Ala-Trp-D-Phe-Lys-NH2)
InChI Formula InChI=1S/C46H56N12O6/c1-27(54-44(62)39(20-29-23-51-35-15-7-5-13-32(29)35)57-43(61)34(48)22-31-25-50-26-53-31)42(60)56-40(21-30-24-52-36-16-8-6-14-33(30)36)46(64)58-38(19-28-11-3-2-4-12-28)45(63)55-37(41(49)59)17-9-10-18-47/h2-8,11-16,23-27,34,37-40,51-52H,9-10,17-22,47-48H2,1H3,(H2,49,59)(H,50,53)(H,54,62)(H,55,63)(H,56,60)(H,57,61)(H,58,64)/t27-,34-,37-,38+,39+,40-/m0/s1
SMILES C[C@H](NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@@H](N)Cc3cnc[nH]3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@H](Cc6ccccc6)C(=O)N[C@@H](CCCCN)C(=O)N
IUPAC Name (2S)-6-amino-2-[[(2R)-2-[[(2S)-2-[[(2S)-2-[[(2R)-2-[[(2S)-2-amino-3-(1H-imidazol-5-yl)propanoyl]amino]-3-(1H-indol-3-yl)propanoyl]amino]propanoyl]amino]-3-(1H-indol-3-yl)propanoyl]amino]-3-phenylpropanoyl]amino]hexanamide
Molecular Weight (g/mol) 873
Quantity 1 mg
Formula Weight 872.4446
Shipping Condition Room Temperature
Chemical Name or Material L-Histidyl-D-tryptophyl-L-alanyl-L-tryptophyl-D-phenylalanyl-L-lysinamide Acetate Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.