missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023866
16.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
18598-74-8
C7H16ClNO2
181.66
MFCD00038911
GGTBEWGOPAFTTH-GEMLJDPKSA-N
l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
11171351
methyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
Cl.CC[C@H](C)[C@H](N)C(=O)OC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
18598-74-8
C7H16ClNO2
181.66
MFCD00038911
GGTBEWGOPAFTTH-GEMLJDPKSA-N
l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
11171351
methyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
Cl.CC[C@H](C)[C@H](N)C(=O)OC

L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

Solubility
Soluble in methanol (50 mg/ml clear, colorless solution).

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. In order to obtain a higher solubility, warm the tube at 37°C and shake it in the ultrasonic bath for a while.
TRUSTED_SUSTAINABILITY
Melting Point 98°C to 100°C
Beilstein 3595132
Solubility Information Soluble in methanol (50mg/ml clear,colorless solution).
Formula Weight 181.66
Percent Purity ≥98%
Quantity 1 g
Sensitivity Hygroscopic
Chemical Name or Material L-Isoleucine methyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'