missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine, TRC

Product Code. 17979264
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979264 100 g 100g
17988224 50 g 50g
17989644 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17979264 Supplier Toronto Research Chemicals Supplier No. TRCI820175100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-32-5
InChI Formula InChI=1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
Molecular Formula C6 H13 N O2
Molecular Weight (g/mol) 131.17
Chemical Name or Material L-Isoleucine

Specifications

Synonym Isoleucine, L-Isoleucine, Isoleucine, L- (8CI), (2S,3S)-2-Amino-3-methylpentanoic acid, (2S,3S)-α-Amino-β-methyl-n-valeric acid, (2S,3S)-α-Amino-β-methylvaleric acid, (S)-Isoleucine, (S,S)-Isoleucine, 2-Amino-3-methylvaleric acid, 2S,3S-Isoleucine, Isoleucine, (2S,3ξ)-, L-(+)-Isoleucine, L-Ile, L-Norvaline, 3-methyl-, erythro-, NSC 46708, Pentanoic acid, 2-amino-3-methyl-, (2S,3S)-, [S-(R*,R*)]-2-Amino-3-methylpentanoic acid, erythro-L-Isoleucine
SMILES CC[C@H](C)[C@H](N)C(=O)O
Quantity 100 g
Formula Weight 131.0946
Percent Purity >95%
Shipping Condition Room Temperature
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.