missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine-d7-N-t-BOC H2O (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | N-(tert-Butoxycarbonyl)-L-leucine-4,5,5,5,5',5',5'-D7 Hydrate |
| Molecular Formula | C11 D7 H14 N O4 . H2 O |
| Quantity | 0.1 g |
| Synonym | (S)-2-((tert-Butoxycarbonyl)amino)-4-(methyl-d3)pentanoic-4,5,5,5-d4 acid hydrate, N-[(1,1-Dimethylethoxy)carbonyl]-L-leucine-d7 hydrate, (2 S)-4-(Methyl-d3)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic-4,5,5,5-d4 acid monohydrate, L-Boc leucine-d7 hydrate, N-(tert-Butoxycarbonyl)-L-leucine-4,5,5,5,5',5',5'-D7 hydrate, L-Leucine-d7-N-t-BOC H2O (iso-propyl-d7), L-Leucine-d7-N-t-BOC H2O (iso-propyl-d7), L-Leucine-d7-N-t-BOC H2O (iso-propyl-d7) |
| InChI Formula | InChI=1 S/C11H21NO4.H2O/c1-7(2)6-8(9(13)14)12-10(15)16-11(3,4)5;/h7-8 H,6H2,1-5H3,(H,12,15)(H,13,14);1H2/t8-;/m0./s1/i1D3,2D3,7 D; |
| SMILES | O.[2 H]C([2 H])([2 H])C([2 H])(C[C@H](NC(=O)OC(C)(C)C)C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | (2 S)-4,5,5,5-tetradeuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-(trideuteriomethyl)pentanoic acid;hydrate |
| Molecular Weight (g/mol) | 256.3472 |
| Formula Weight | 256.2016 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction