Learn More
L-Menthyl Acetate, TRC
Product Code. 30138259
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
Unit Size:
10mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138259 10 mL 10mL
30004891 5 mL 5mL
30007939 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138259 Supplier Toronto Research Chemicals Supplier No. TRCM21915010ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2623-23-6
Molecular Formula C12 H22 O2
Molecular Weight (g/mol) 198.3
Synonym (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol 1-Acetate, [1R-(1α,2β,5α)]-5-Methyl-2-(1-methylethyl)cyclohexanol Acetate, (1R,3R,4S)-(-)-Menthol Acetate, (-)-Menthol Acetate, (-)-Menthyl Acetate, (-)-p-Menthyl Acetate, (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol Acetate, Acetic Acid (R)-Menthyl Ester, NSC 3722, NSC 52970, l-(-)-Menthyl Acetate, l-Menthol Acetate
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] acetate
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)C
Specifications
Quantity 10 mL
InChI Formula InChI=1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12-/m1/s1
Formula Weight 198.162
Chemical Name or Material L-Menthyl Acetate