missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine, MP Biomedicals™

Product Code. 11403520 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403520 100 g 100g
11493510 25 g 25g
11413520 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403520 Supplier MP Biomedicals Supplier No. 02102291.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Methionine, MP Biomedicals is one of the common sulfur-containing amino acids. It is a methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol and choline.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 284°C (decomposition)(literature)
pH 5.6 to 6.1 (1% aq. soln.)
Quantity 100 g
Solubility Information Soluble in warm dil alcohol; insoluble in abs. alcohol, ether, petroleum ether, benzene, acetone, HCl.
Optical Rotation 22.4–24.7° (c=20, 6N HCl) (Lit.)
Formula Weight 149.21
Percent Purity ≥99%
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.