missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals

Catalog Number p-3776601
Click to view available options
Quantity:
1 g
5 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
327-57-1
C6H13NO2
131.18
MFCD00064423
LRQKBLKVPFOOQJ-YFKPBYRVSA-N
l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate
21236
CHEBI:18347
(2S)-2-aminohexanoic acid
CCCC[C@H](N)C(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
327-57-1
C6H13NO2
131.18
MFCD00064423
LRQKBLKVPFOOQJ-YFKPBYRVSA-N
l-norleucine, norleucine, h-nle-oh, s-2-aminohexanoic acid, caprine, glycoleucine, l-+-norleucine, l-2-aminohexanoic acid, 2-aminocaproic acid, l-2-aminohexanoate
21236
CHEBI:18347
(2S)-2-aminohexanoic acid
CCCC[C@H](N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White
Assay Percent Range 99+%
Infrared Spectrum Authentic
Beilstein 04, 432
Loss on Drying 1% max. (105°C, 4 hrs)
Merck Index 15, 6795
Other Amino Acids (TLC) not detectable
Packaging Glass Bottle
Solubility Information Solubility in water: 1.6 g/100mL (23 c).
Specific Rotation + 22.00
Formula Weight 131.17
Percent Purity ≥99%
Sulfated Ash 0.5% max.
Sublimation Point 0275.0°C
Specific Rotation Condition +22° (20°C c=2,6N HCl)
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Norleucine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'