missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Prolyl-L-isoleucine, TRC
Specifications
Specifications
| Chemical Name or Material | L-Prolyl-L-isoleucine |
| CAS | 51926-51-3 |
| Molecular Formula | C11 H20 N2 O3 |
| Quantity | 5 mg |
| Synonym | L-Isoleucine','L-prolyl- (9CI','ACI), L-Isoleucine','N-L-prolyl- (ZCI), L-Prolyl-L-isoleucine (ACI), 168: PN: WO2011146121 PAGE: 117 claimed sequence, 19: PN: WO2010099019 SEQID: 6 unclaimed sequence, 2003: PN: EP2071334 SEQID: 2112 claimed protein, 2003: PN: WO2009077864 SEQID: 2112 claimed protein, 377: PN: EP2161028 PAGE: 14 claimed protein, 452: PN: WO2005081628 SEQID: 454 claimed protein, 472: PN: WO2022125673 SEQID: 472 claimed protein, 480: PN: WO2022215055 SEQID: 480 claimed protein, 53: PN: WO2021211467 SEQID: 206 claimed sequence |
| InChI Formula | InChI=1S/C11H20N2O3/c1-3-7(2)9(11(15)16)13-10(14)8-5-4-6-12-8/h7-9,12H,3-6H2,1-2H3,(H,13,14)(H,15,16)/t7-,8-,9-/m0/s1 |
| SMILES | CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1)C(=O)O |
| IUPAC Name | (2S,3S)-3-methyl-2-[[(2S)-pyrrolidine-2-carbonyl]amino]pentanoic acid |
| Molecular Weight (g/mol) | 228.288 |
| Formula Weight | 228.1474 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?