missing translation for 'onlineSavingsMsg'
Learn More

L-Prolyl-L-isoleucine, TRC

Product Code. 30409285
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409285 Supplier Toronto Research Chemicals Supplier No. TRCP83997350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Prolyl-L-isoleucine
CAS 51926-51-3
Molecular Formula C11 H20 N2 O3
Quantity 50 mg
Synonym L-Isoleucine','L-prolyl- (9CI','ACI), L-Isoleucine','N-L-prolyl- (ZCI), L-Prolyl-L-isoleucine (ACI), 168: PN: WO2011146121 PAGE: 117 claimed sequence, 19: PN: WO2010099019 SEQID: 6 unclaimed sequence, 2003: PN: EP2071334 SEQID: 2112 claimed protein, 2003: PN: WO2009077864 SEQID: 2112 claimed protein, 377: PN: EP2161028 PAGE: 14 claimed protein, 452: PN: WO2005081628 SEQID: 454 claimed protein, 472: PN: WO2022125673 SEQID: 472 claimed protein, 480: PN: WO2022215055 SEQID: 480 claimed protein, 53: PN: WO2021211467 SEQID: 206 claimed sequence
InChI Formula InChI=1S/C11H20N2O3/c1-3-7(2)9(11(15)16)13-10(14)8-5-4-6-12-8/h7-9,12H,3-6H2,1-2H3,(H,13,14)(H,15,16)/t7-,8-,9-/m0/s1
SMILES CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1)C(=O)O
IUPAC Name (2S,3S)-3-methyl-2-[[(2S)-pyrrolidine-2-carbonyl]amino]pentanoic acid
Molecular Weight (g/mol) 228.288
Formula Weight 228.1474
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.