Learn More
L-Rhamnose Diethyl Dithioacetal, TRC
Specifications
Specifications
| Chemical Name or Material | L-Rhamnose Diethyl Dithioacetal |
| CAS | 6748-70-5 |
| Molecular Formula | C10 H22 O4 S2 |
| Quantity | 50 g |
| InChI Formula | InChI=1S/C10H22O4S2/c1-4-15-10(16-5-2)9(14)8(13)7(12)6(3)11/h6-14H,4-5H2,1-3H3/t6-,7-,8+,9+/m0/s1 |
| SMILES | CCSC(SCC)[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O |
| IUPAC Name | (2R,3R,4S,5S)-1,1-bis(ethylsulfanyl)hexane-2,3,4,5-tetrol |
| Molecular Weight (g/mol) | 270.41 |
| Formula Weight | 270.096 |