Learn More
Masterflex™ L/S™ Bulk-Packed Platinum-Cured Silicone High-Performance Precision Pump Tubing
Click to view available options
Tubing Size:
13
14
15
16
17
24
25
35
36
Flow Rate:
1000 mL/min
1100mL/min.
130 mL/min
1700 mL/min
1800 mL/min
2600 mL/min
3400 mL/min
36 mL/min
480 mL/min
Unit Size:
Pack of 10
Pack of 20
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Slightly greater clarity than peroxide-cured tubing
- Smooth surfaces; lower protein binding levels
- Fewer potential leachables
- Pharmaceutical, biotech, peristaltic pumping, and food applications
- Excellent heat resistance, low compression set, odorless, nontoxic, translucent
- Comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size).
- Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping.
- Tubing comes in 25-ft bags.
Specifications
Specifications
| Material | Platinum-cured Silicone |
| Overall Length (English) | 25 ft. |
| Overall Length (Metric) | 7.6 m |
| For Use With (Application) | Pharmaceutical, Biotech, Peristaltic Pumping, and Food Applications |
| Min. Operating Temperature (Metric) | -40°C |
| Tubing Size | 17 |
| Flow Rate | 1700 mL/min |
| Hose Barb | 1/4 in. |
| Max. Operating Temperature (Metric) | 100°C |