missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Selectin/CD62L Antibody (95218), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals FAB5761H
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
| L-Selectin/CD62L | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 95218 | |
| Flow Cytometry, CyTOF-ready | |
| CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
| Mouse myeloma cell line NS0-derived recombinant mouse L-Selectin/CD62L, Met1-Asn332, Accession # P18337 | |
| 0.5 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cardiovascular Biology, Glycobiology, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cells | |
| 6402 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction