Learn More
Techsit™ L-Tech Laboratory Saddle Seat
Click to view available options
Color:
Black
Sapphire Blue
Unit Size:
Each
Description
- Vinyl saddle stool on casters
- Easy-to-clean antimicrobial vinyl
- Fully adjustable seat height and seat tilt
- Ceanroom Class 7 suitable
Specifications
Specifications
| Product Type | Saddle Seat |
| Color | Sapphire Blue |
| Adjustable | Yes |
| Finish | Vinyl |
| Frame Material | Steel |
| Lift Type | Gas Lift |
| Load Bearing Capacity (Metric) | 150 kg |
| Seating Height | 420 to 530 mm |
| Upholstery | Vinyl |
| Warranty | 5 Year |
| Show More |