Learn More
L-Tetrahydrofolic Acid (>70% when packaged), TRC
Specifications
Specifications
| Chemical Name or Material | L-Tetrahydrofolic Acid |
| CAS | 135-16-0 |
| Molecular Formula | C19 H23 N7 O6 |
| Quantity | 10 mg |
| Synonym | L-Glutamic acid','N-[4-[[(2-amino-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]- (ACI), L-Glutamic acid','N-[4-[[(2-amino-1,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]- (9CI), Glutamic acid','N-[p-[[(2-amino-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-','L- (7CI','8CI), N-[4-[[(2-Amino-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid (ACI), (-)-L-5,6,7,8-Tetrahydrofolic acid, (2S)-2-(4-(((2-Imino-4-oxo-1,2,3,4,5,6,7,8-octahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, 5,6,7,8-Tetrahydrofolic acid, L-5,6,7,8-Tetrahydrofolic acid, Tetrahydrofolic acid, Tetrahydropteroylglutamic acid, THFA, L-Tetrahydrofolic Acid |
| InChI Formula | InChI=1S/C19H23N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,11-12,21,23H,5-8H2,(H,24,29)(H,27,28)(H,31,32)(H4,20,22,25,26,30)/t11?,12-/m0/s1 |
| SMILES | NC1=NC(=O)C2=C(NCC(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)N2)N1 |
| IUPAC Name | (2S)-2-[[4-[(2-amino-4-oxo-5,6,7,8-tetrahydro-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid |
| Molecular Weight (g/mol) | 445.43 |
| Formula Weight | 445.171 |