missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophan benzyl ester hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
L-Tryptophan benzyl ester hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol (50 mg/mL in MeOH).
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
L-Tryptophan benzyl ester hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in methanol (50 mg/mL in MeOH).
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Color | White |
Solubility Information | Soluble in methanol (50mg/mL in MeOH). |
IUPAC Name | benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl |
Formula Weight | 330.81 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | L-Tryptophan benzyl ester hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'