Learn More
L-Tryptophan-d3, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | L-Tryptophan-d3 |
| CAS | 133519-78-5 |
| Molecular Formula | C11 D3 H9 N2 O2 |
| Quantity | 25 mg |
| Synonym | L-Tryptophan-α,β,β-d3 (9CI), α,β,β-Tri-2H-L-tryptophan, L-Tryptophan-α,β,β-d3, L-Tryptophan-d3, (S)-α-Amino-3-indole-α,β,β-d3-propionic acid, (αS)-α-Aminoindole-3-propanoic-d3 acid, L-Tryptophan-α,β,β-D3 |
| InChI Formula | InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1/i5D2,9D |
| SMILES | [2H]C([2H])(c1c[nH]c2ccccc12)[C@]([2H])(N)C(=O)O |
| IUPAC Name | (2S)-2-amino-2,3,3-trideuterio-3-(1H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 207.244 |
| Formula Weight | 207.109 |