missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1/ORF2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-44223-0.05MG
This item is not returnable.
View return policy
Description
L1/ORF2 Polyclonal specifically detects L1/ORF2 in Mouse samples. It is validated for ELISA.
Specifications
| L1/ORF2 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
| Chicken | |
| 0.05 mg | |
| Mouse | |
| IgG |
| ELISA | |
| 1.21 mg/mL | |
| ELISA 1:20000-1:100000 | |
| P11369 | |
| This IgY fraction antibody was prepared from eggs of chickens laid after repeated immunizations with three synthetic peptides conjugated to keyhole limpet hemocyanin (KLH). The peptides correspond to regions within the endonuclease domain of L1/ORF2 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction