Learn More
L1CAM Antibody (CL12988) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18553AF594
Product Code. 30290752
  • € 780.00 / 0.10mL
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
L1CAM
Monoclonal
Alexa Fluor 594
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
This antibody was developed using a synthetic peptide derived from P32004, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
3897
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12988
50mM Sodium Borate
Mouse
Protein A purified