missing translation for 'onlineSavingsMsg'
Learn More

L1CAM Antibody (CL12988), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-18553-25ul

 View more versions of this product

Product Code. 18688654

  • € 429.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

L1CAM
Monoclonal
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12988
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
This antibody was developed against a sythetic peptide for P32004. Actual immunogen sequence is proprietary.
25 μg
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
3897
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.