missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (L1CAM/9267R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24129CL1
This item is not returnable.
View return policy
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| L1CAM | |
| Monoclonal | |
| CoraFluor 1 | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Recombinant fragment (around aa600-900) of human L1CAM protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| 3897 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| L1CAM/9267R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction