missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (rL1CAM/9163), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | L1CAM |
|---|---|
| Clone | rL1CAM/9163 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683024
|
Novus Biologicals
NBP3-23680-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673464
|
Novus Biologicals
NBP3-23680-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| L1CAM | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 3897 | |
| IgG2b κ | |
| Protein A or G purified |
| rL1CAM/9163 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Recombinant full-length human L1CAM protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title