missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (UJ181.4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2683H
This item is not returnable.
View return policy
Description
L1CAM Monoclonal specifically detects L1CAM in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| L1CAM | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| L1CAM | |
| 16 week human foetal brain. | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuroscience | |
| 3897 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| UJ181.4 | |
| Western Blot, Immunohistochemistry | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction