missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501437
This item is not returnable.
View return policy
Description
L1CAM (CD171) is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant.Specifications
| L1CAM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P32004 | |
| L1CAM | |
| Full length human recombit protein of human L1CAM produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1A8 | |
| Unconjugated | |
| L1CAM | |
| CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3897 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction