missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ L1RE1 Recombinant Rabbit Monoclonal Antibody (K01_2N11)
GREENER_CHOICE

Product Code. 30824805
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824805 100 μL
30828808 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824805 Supplier Invitrogen™ Supplier No. MA564112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

L1RE1 Recombinant Monoclonal Antibody for Western Blot

L1RE1 encodes a nucleic acid-binding protein, which is essential for retrotransposition of LINE-1 elements in the genome. The proteins act as nucleic acid chaperones binding its own transcript and therefore preferentially mobilizing the transcript from which they are encoded. Long interspersed element-1/LINE-1/L1 retrotransposons are present in more than 500'000 full (6 kb) or truncated copies in the human genome. Most of them are inactive but one estimate is that 80 to 100 of those elements could be transcribed, translated and active in any individual. An active LINE-1 encodes for 2 proteins translated from a single RNA containing two non-overlapping ORFs, ORF1 and ORF2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L1RE1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2N11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene L1RE1
Gene Accession No. Q9UN81
Gene Symbols L1RE1
Host Species Rabbit
Immunogen Recombinant protein of human LINE-1 ORF1p.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4029
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.