missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1TD1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81960AF750
This item is not returnable.
View return policy
Description
L1TD1 Polyclonal antibody specifically detects L1TD1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| L1TD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 54596 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 750 | |
| ECAT11, ES cell-associated protein 11, FLJ10884, LINE-1 type transposase domain containing 1, LINE-1 type transposase domain-containing protein 1, MGC133253, RP5-1155K23.3 | |
| L1TD1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human L1TD1. The immunogen is located within amino acids 690 - 740 of L1TD1. Amino Acid Squence: IPEKESYENRAEDIIKE | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction