missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1TD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21528-1-AP-150UL
This item is not returnable.
View return policy
Description
L1TD1 is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.Specifications
| L1TD1 | |
| Polyclonal | |
| Unconjugated | |
| L1TD1 | |
| ECAT11, ES cell associated protein 11, L1TD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54596 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5T7N2 | |
| L1TD1 | |
| L1TD1 Fusion Protein Ag16147 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction