missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47314H
This item is not returnable.
View return policy
Description
L3MBTL3 Polyclonal antibody specifically detects L3MBTL3 in Human samples. It is validated for Western Blot
Specifications
| L3MBTL3 | |
| Polyclonal | |
| Western Blot | |
| H-l(3)mbt-like protein 3, KIAA1798, l(3)mbt-like 3 (Drosophila), L(3)mbt-like protein 3, lethal(3)malignant brain tumor-like protein 3, MBT1, MBT-1, RP11-73O6.1 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Lethal(3)Malignant Brain Tumor-like 3 using the numbering given in entry NP_115814.1 (GeneID 84456). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84456 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction