missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL3 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00084456-B01P
This item is not returnable.
View return policy
Description
L3MBTL3 Polyclonal antibody specifically detects L3MBTL3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| L3MBTL3 | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence | |
| AAH60845.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 84456 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| H-l(3)mbt-like protein 3, KIAA1798, l(3)mbt-like 3 (Drosophila), L(3)mbt-like protein 3, lethal(3)malignant brain tumor-like protein 3, MBT1, MBT-1, RP11-73O6.1 | |
| L3MBTL3 (AAH60845.1, 1 a.a. - 755 a.a.) full-length human protein. MTESASSTSGQEFDVFSVMDWKDGVGTLPGSDLKFRVNEFGALEVITDENEMENVKKATATTTWMVPTAQEVFSEKTGMPFRLKDPVKVEGLQFCENCCQYGNVDECLSGGNYCSQNCARHIKDKDQKEERDVEEDNEEEDPKCSRKKKPKLSLKADNKEDGEERDDEMENKQDVRILRGSQRARRKRRGDSAVLKQGLPPKGKKAWCWASYLEEEKAVAVPAKLFKEHQSFPYNKNGFKVGMKLEGVDPEHQSVYCVLTVAEVCGYRIKLHFDGYSDCYDFWVNADALDIHPVGWCEKTGHKLHPPKGYKEEEFNWQTYLKTCKAQAAPKSLFENQNITVIPSGFRVGMKLEAVDKKNPSFICVATVTDMVDNRFLVHFDNWDESYDYWCEASSPHIHPVGWCKEHRRTLITPPGYPNVKHFSWDKYLEETNSLPAPARAFKVKPPHGFQKKMKLEVVDKRNPMFIRVATVADTDDHRVKVHFDGWNNCYDYWIDADSPDIHPVGWCSKTGHPLQPPLSPLELMEASEHGGCSTPGCKGIGHFKRARHLGPHSAANCPYSEINLNKDRIFPDRLSGEMPPASPSFPRNKRTDANESSSSPEIRDQHADDVKEDFEERTESEMRTSHEARGAREEPTVQQAQRRSAVFLSFKSPIPCLPLRWEQQSKLLPTVAGIPASKVSKWSTDEVSEFIQSLPGCEEHGKVFKDEQIDGEAFLLMTQTDIVKIMSIKLGPALKIFNSILMFKAAEKNSHNEL | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction