missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL3 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809709
This item is not returnable.
View return policy
Description
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.Specifications
| L3MBTL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96JM7 | |
| L3MBTL3 | |
| Human recombit protein fragment corresponding to amino acids 98-319 of human L3MBTL3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1H2 | |
| Unconjugated | |
| L3MBTL3 | |
| MBT-1, MBT1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84456 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction