missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL3 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809709S
This item is not returnable.
View return policy
Description
L3MBTL3 Monoclonal antibody specifically detects L3MBTL3 in Human, Mouse samples. It is validated for Western BlotSpecifications
| L3MBTL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P09017, Q08624 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 15423, 3221 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1H2 | |
| Unconjugated | |
| HOXC4 | |
| AI481284; H-l(3)mbt-like protein 3; KIAA1798; l(3)mbt-like 3; l(3)mbt-like 3 (Drosophila); l(3)mbt-like protein 3; L3MBTL histone methyl-lysine binding protein 3; L3mbtl3; L3MBTL3, histone methyl-lysine binding protein; lethal(3)malignant brain tumor-like protein 3; lethal(3)malignant brain tumor-like protein 3; LOW QUALITY PROTEIN: lethal(3)malignant brain tumor-like protein 3; MBT1; MBT-1 | |
| Full length human recombinant protein of human HOXC4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction