missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L3MBTL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584139
This item is not returnable.
View return policy
Description
Immunogen sequence: SEKTGMPFRL KDPVKVEGLQ FCENCCQYGN VDECLSGGNY CSQNCARHIK DKDQKEERDV EEDNEEEDPK CSRKK Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 83%.
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
Specifications
| L3MBTL3 | |
| Polyclonal | |
| Unconjugated | |
| L3MBTL3 | |
| AI481284; H-l(3)mbt-like protein 3; KIAA1798; l(3)mbt-like 3; l(3)mbt-like 3 (Drosophila); l(3)mbt-like protein 3; L3MBTL histone methyl-lysine binding protein 3; L3mbtl3; L3MBTL3, histone methyl-lysine binding protein; lethal(3)malignant brain tumor-like protein 3; lethal(3)malignant brain tumor-like protein 3; LOW QUALITY PROTEIN: lethal(3)malignant brain tumor-like protein 3; MBT1; MBT-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84456 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JM7 | |
| L3MBTL3 | |
| Recombinant protein corresponding to Human L3MBTL3. Recombinant protein control fragment (Product #RP-93657). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction