missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00091133-D01P
This item is not returnable.
View return policy
Description
L3MBTL4 Polyclonal antibody specifically detects L3MBTL4 in Human samples. It is validated for Western Blot
Specifications
| L3MBTL4 | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_775735.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| FLJ35936, H-l(3)mbt-like protein 4, HsT1031, l(3)mbt-like 4 (Drosophila), L(3)mbt-like protein 4, lethal(3)malignant brain tumor-like protein 4 | |
| L3MBTL4 (NP_775735.1, 1 a.a. - 534 a.a.) full-length human protein. MKQPNRKRKLNMDSKERLDQDGRLEQAEEEKKPKDSTTPLSHVPSAAAQGAWSWEWYLKEQKAVAAPVELFSKDQSFPEHENGFQIGMRLEGIDPRHPSVFCVLSVAEVCGYRLRLHFDGYLSCYDFWTNAGSPDIHPVGWCEKTKHELHIPKGYRKDKFVWMDYLKACKLQNAPKKLFRNRSPNGPMSKEFQVGMKLEAVDRKNPSLVCVATIADIVEDRLLVHFDNWDDSYDYWCDVNSPYVQPVGWCQENGRTLIAPQGYPNPENFSWTEYLEATQTNAVPAKVFKMRLPHGFLPNMKLEVVDKRNPRLIRVATIVDVDDQRVKVHFDGWDHKYDYWVEADSPDIHPIGWCDVTGHPLEVPQRTNDLKILPGQAVCPTPGCRGIGHIRGPRYSGHHSAFGCPYSDMNLKKEATLHDRLREQTQANLESDSSHSKSKSLCSLNFNGKHEKVNSQPRLVQQAKCLKIKGKEDIDLDNLFRGRVRWLMPVIPALWEAEAGRSRDQEIKTILANAVKPRLYQKYKKLARRGGGCL | |
| 0.1 mg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 91133 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction