missing translation for 'onlineSavingsMsg'
Learn More
Learn More
La Crosse Virus Glycoprotein 1 Antibody (807.31), Alexa Fluor™ 594 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20121AF594
This item is not returnable.
View return policy
Description
La Crosse Virus Glycoprotein 1 Monoclonal antibody specifically detects La Crosse Virus Glycoprotein 1 in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| La Crosse Virus Glycoprotein 1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Bunyavirus La Crosse Glycoprotein 1, G1 envelope glycoprotein, Glycoprotein C, La Crosse Virus Gc, LCV G1, M polyprotein G1 chain | |
| This antibody was raised by immunising BALB/c mice with La Crosse viruses and the harvested spleens were fused with mouse P3X63Ag8.653 myeloma cells. | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 807.31 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 956555 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction