missing translation for 'onlineSavingsMsg'
Learn More
Learn More
La Crosse Virus Glycoprotein 1 Antibody (807.31) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20122-0.2mg
This item is not returnable.
View return policy
Description
La Crosse Virus Glycoprotein 1 Monoclonal antibody specifically detects La Crosse Virus Glycoprotein 1 in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Neutralization
Specifications
| La Crosse Virus Glycoprotein 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 956555 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Neutralization | |
| 807.31 | |
| Unconjugated | |
| Bunyavirus La Crosse Glycoprotein 1, G1 envelope glycoprotein, Glycoprotein C, La Crosse Virus Gc, LCV G1, M polyprotein G1 chain | |
| This antibody was raised by immunising BALB/c mice with La Crosse viruses and the harvested spleens were fused with mouse P3X63Ag8.653 myeloma cells. | |
| 0.2 mg | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction