missing translation for 'onlineSavingsMsg'
Learn More
Learn More
La Crosse Virus Nucleocapsid Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41225JF646
This item is not returnable.
View return policy
Description
La Crosse Virus Nucleocapsid Polyclonal antibody specifically detects La Crosse Virus Nucleocapsid in Virus samples. It is validated for ELISA
Specifications
| La Crosse Virus Nucleocapsid | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| Janelia Fluor 646 | |
| LACVsSgp1, N, Nucleocapsid protein, Nucleoprotein, Protein N | |
| Antibody was raised against an 18 amino acid peptide near the center of the La Crosse virus nucleocapsid. The immunogen is located within amino acids 90 - 140 of La Crosse Virus Nucleocapsid. Amino Acid Squence: LDQYNENDDESQHELIRT | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 956556 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction