Learn More
Actin, Smooth Muscle, Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody.
€ 112.20 - € 495.00
Specifications
Antigen | Actin, Smooth Muscle Ab-1 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Regulatory Status | IVD |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12669076
|
Epredia
MS-113-B1 |
Concentrated | Biotin | 500 μL |
€ 435.00
500µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12699076
|
Epredia
MS-113-P1 |
Concentrated | Unconjugated | 500 μL |
€ 329.00
500µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12679076
|
Epredia
MS-113-P |
Concentrated | Unconjugated | 1 mL |
€ 408.00
1mL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12649086
|
Epredia
MS-113-R7 |
Prediluted | Unconjugated | 7 mL |
€ 188.00
7mL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12659086
|
Epredia
MS-113-RQ |
Ready-to-Use | Unconjugated | 12 mL |
€ 283.00
12mL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12619086
|
Epredia
MS-113-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12659076
|
Epredia
MS-113-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12649076
|
Epredia
MS-113-B |
Concentrated | Biotin | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12689076
|
Epredia
MS-113-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12609086
|
Epredia
MS-113-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Host Species: Mouse
Clone: 1A4; also known as asm-1
Isotype: IgG2a/κ
Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Epitope: Acetyl group and the first four amino acids on the N-terminal end of the peptidic chain of α-smooth actin
Immunogen: N-terminal decapeptide of α-smooth muscle isoform of actin; acetylated at the N-terminus
Positive Control: Smooth muscle or leiomyosarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Actin, Smooth Muscle Ab-1 | |
Monoclonal | |
IVD | |
N-terminal decapeptide of alpha-smooth muscle isoform of actin; acetylated at the N-terminus |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Mouse | |
Muscle Biology | |
Primary |