Learn More
Cytokeratin 10 Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
Brand: Epredia MS-611-P
Additional Details : Weight : 4.98950kg
Description
CK10 (Cytokeratin 10) is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.Host Species: Mouse
Clone: DE-K10
Isotype: IgG1
Species Reactivity: Human, Rat, Dog and Cat. Others not known.
Epitope: Not determined
Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium
Molecular Weight: 56.5 kDa
Positive Control: A431 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Cytokeratin 10 Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cytoskeletal Protein | |
Canine, Feline, Human, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
DE-K10 | |
Unconjugated | |
Cytoskeletal preparation extracted from human ectocervical epithelium | |
IVD | |
Primary | |
Concentrated |