missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody

Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ MS-799-P1ABX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12632327

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: A103

Isotype: IgG1

Species Reactivity: Human and Dog. Others not tested.

Epitope: Not determined

Immunogen: Recombinant MART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and Melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
Specifications

Specifications

MART-1/Melan-A (Melanoma Marker) Ab-4
Monoclonal
1mg/mL
Mouse
100 μL
Cancer and Tumor Biology
Canine, Human
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
A103
Unconjugated
Recombinant MART-1 protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers