missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available wit the Epredia™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-364-S0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12691187

  • € 154.00 / 100µL
Estimated Shipment: 16-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Research Areas: Cancer Marker

Host Species: Mouse

Clone: HMB45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with dog and rat. Others not known.

Immunogen: Extract of pigmented melanoma metastases from lymph nodes

Molecular Weight: 100kDa

Positive Control: Melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

    Western Blotting

Specifications

Specifications

Melanoma (gp100) Ab-1
Monoclonal
Unconjugated
Extract of pigmented melanoma metastases from lymph nodes
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin), Western Blot
HMB45
Mouse
100 μL
Cancer and Tumor Biology
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers