Learn More
Progesterone Receptor Ab-8 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.
Brand:  Epredia MS-298-PABX
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12690867
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.
Host Species: Mouse
Clone: hPRa 2 + hPRa 3
Isotype: IgG1/κ + IgG1/κ
Species Reactivity: Human, Horse, Sheep and Pig. Others not known.
Epitope: N-terminal half of human PgR
Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (B-form) and 81kDa (A-form)
Positive Control: T47D or Breast CA
Cellular Localization: Nuclear
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient
Specifications
Specifications
Progesterone Receptor Ab-8
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Equine, Human, Porcine, Sheep
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
hPRa 2+hPRa 3
Unconjugated
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
ASR, IVD
Primary
Concentrated