Learn More
Smooth Muscle Actin Rabbit Polyclonal Antibody, Epredia™
Stain smooth muscle cells in vessel walls, gut wall, and myometrium with Epredia™ Actin, Smooth Muscle, Rabbit Polyclonal Antibody.
Brand: Epredia RB-9010-P
Additional Details : Weight : 0.00250kg
Description
This antibody stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not tested.
Epitope: N-terminus
Immunogen: A synthetic peptide from N-terminus of human α-smooth muscle isoform of actin
Positive Control: Smooth muscle of arteries, gut wall, uterus and Leiomyoma
Cellular Localization: Cytoplasmic.
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Actin, Smooth Muscle | |
Polyclonal | |
Unconjugated | |
A synthetic peptide from N-terminus of human alpha-smooth muscle isoform of actin | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
200µg/mL | |
Rabbit | |
1 mL | |
Muscle Biology | |
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit | |
IgG |