Learn More
Labconco™ 48 Port Manifold Gasket
Product Code. 15859674 Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
For 48 port manifold
Unit Size:
Each
Product Code. 15859674
Brand: Labconco™ 7607300
Only null left
Add to basket
Add to basket
Replacement port gasket creates air-tight seal between FreeZone Freeze Dryer and main chamber.
  • Replacement port gasket
  • Sits under main chamber
  • Creates air-tight seal
  • For Labconco FreeZone Freeze Dryers
Specifications
For Use With (Equipment) For 48 port manifold
Diameter (English) Outside 4.5 in.
Diameter (Metric) Outside 11.43 cm
Material Neoprene
Type Gasket, Bottom