Learn More
Labconco™ CentriVap™ Strobe Light for CentriVap™ System
Product Code. 10175133 Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
CentriVap Centrifugal System
Unit Size:
Each
Product Code. 10175133
Brand: Labconco™ 7464300
Only null left
Add to basket
Add to basket
Includes
Mounting holster and hardware for attachment to the right side of the CentriVap concentrator
Enable user to view samples while rotor is spinning.
Enable user to view samples while rotor is spinning
  • Makes samples appear to stand still while rotor is in motion and chamber is under vacuum
Specifications
Type Strobe Light
For Use With (Equipment) CentriVap Centrifugal System
Weight (English) 2 lb.
Weight (Metric) 0.9 kg