Learn More
Labconco™ Flask Holder
Shop All Labconco Corporation Products
Click to view available options
Size:
Small
Large
Unit Size:
Each
Description
- Made of stainless-steel
- Allows a small fast-freeze flask bottom containing the sample to be placed inside a larger fast-freeze flask
Specifications
Specifications
| Type | Flask Holder |
| No. of Shelves | 0 |
| Material | Stainless Steel |
| Size | Large |
| For Use With (Application) | To prevent sample melt back |
| For Use With (Equipment) | For fast freeze™ flasks, inner flasks 7542000 (40ml) and 7542200 (80ml) and outer flasks 75411 |
| Width (English) | 3.45 in. |
| Height (English) | 5 in. |
| Height (Metric) | 12.7 cm |
| Weight (English) | 1 lb. |
| Show More |